Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-34 Quantikine ELISA Kit
SDP

Catalog No. M3400 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3400 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M3400 Supplier R&D Systems™ Supplier No. M3400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse IL-34 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-34 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-34 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).

Specifications

Assay Range 39.10 - 2500pg/mL
Assay Sensitivity 15.6pg/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Mouse IL-34.
For Use With (Application) ELISA
Gene ID (Entrez) 76527
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym C16orf77, IL34, IL-34, interleukin 34
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.