Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-38/IL-1F10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067958
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7774RSP 25 μg
MAB7774R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB7774RSP Supplier R&D Systems Supplier No. MAB7774RSP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-38/IL-1F10 Monoclonal specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen IL-38/IL-1F10
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 798036R
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8R459
Gene Alias FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Gene Symbols Il1f10
Host Species Rat
Immunogen E.coli-derived recombinant mouse IL-38/IL-1F10 Met1-Arg152 Accession # Q8R459
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84639
Test Specificity Detects mouse IL-38/IL-1F10 in direct ELISAs and Western blots.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.