Learn More
Abcam Mouse IL-6 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2225031X96TESTS
Encompass_Preferred
Mouse IL-6 ELISA kit is a single-wash 90-min SimpleStep ELISA used to quantification of mouse interleukin-6 (IL-6) with a sensitivity of 11.3 pg/mL. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450nm readout; works on any plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well for higher throughput - Broad sample compatibility - works with serum, plasma, and cell culture supernatants - Cited in over 130 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2050349
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.