Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-6/IL-6R alpha Protein Chimera

Catalog No. 9038SR025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-38S-R025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-38S-R025 Supplier R&D Systems™ Supplier No. 9038SR025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-6/IL-6R alpha Protein Chimera is derived from NS0. The Recombinant Mouse IL-6/IL-6R alpha Protein Chimera has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Molecular Weight (g/mol) Observed M.W.: 70-87kDa, reducing conditions, Theoretical M.W.: 60kDa
Name IL-6/IL-6R alpha Complex
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse IL-6/IL-6R alpha Complex protein, (N-terminus) Mouse IL-6R alpha (Leu20-Glu357) Accession &Num; P22272 GGGSGGGSGGGS Mouse IL-6 (Phe25-Thr211) Accession &Num; P08505 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name IL-6/IL-6 R alpha Complex
Gene Symbol Il6
Biological Activity Measured by its ability to inhibit M1 mouse myeloid leukemia cell proliferation. The ED50 for this effect is typically 2-12ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Mouse IL-6 R alpha (Leu20-Glu357) Accession # P22272, N-terminus; GGGSGGGSGGGS; Mouse IL-6 (Phe25-Thr211) Accession # P08505, C-terminus
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.