Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-6 Quantikine ELISA Kit

Catalog No. M6000B
Change view
Click to view available options
Quantity:
1 Kit (2 Plates)
1 Pk. (6 Plates)
Sufficient For:
2 x 96-well Plates
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
M6000B 1 Kit (2 Plates) 2 x 96-well Plates
SM6000B 1 Pk. (6 Plates) 6 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. M6000B Supplier R&D Systems™ Supplier No. M6000B

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Mouse IL-6 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-6 Quantikine ELISA Kit quantifies mouse IL-6 in cell culture supernates (50 ul), serum (50 ul), heparin plasma (50 ul).

Specifications

Assay Range 7.80 to 500 pg/mL
Assay Sensitivity 1.8 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (Heparin), Plasma (Heparin), Serum, Serum
Specificity Natural and Recombinant Mouse IL-6
Sufficient For 2 x 96-well Plates
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 3569
Gene Symbol Il6
Quantity 1 Kit (2 Plates)
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),Heparin Plasma (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2 A
Target IL-6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.