Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-7R alpha/CD127 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Catalog No. FAB47742N
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB47742N 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB47742N Supplier R&D Systems Supplier No. FAB47742N
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-7R alpha/CD127
Applications Flow Cytometry
Classification Monoclonal
Clone A7R34
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Gene Symbols IL7R
Host Species Rat
Immunogen Recombinant mouse IL-7 R alpha/CD127
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3575
Test Specificity Detects mouse IL-7 R alpha/CD127 in flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.