Learn More
Sino Biological IL13RA2 / CD213A2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 50061RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL13RA2 (Catalog#50061-M08H; NP_032382.1; Met 1-Lys 334). IL13RA2 specific IgG was purified by human IL13RA2 affinity chromatography.
Catalog No. 50-255-3500
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.