Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Il1a (P01582, 115 a.a. - 270 a.a.) Partial Recombinant Protein

Catalog No. 89948735 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-735 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-735 Supplier Abnova™ Supplier No. P4356
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Sequence: SAPYTYQSDLRYKLMKLVRQKFVMNDSLNQTIYQDVDKHYLSTTWLNDLQQEVKFDMYAYSSGGDDSKYPVTLKISDSQLFVSAQGEDQPVLLKELPETPKLITGSETDLIFFWKSINSKNYFTSAAYPELFIATKEQSRVHLARGLPSMTDFQIS

Specifications

Accession Number P01582
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 16175
Molecular Weight (g/mol) 18kDa
Name Il1a (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 10 μg
Immunogen SAPYTYQSDLRYKLMKLVRQKFVMNDSLNQTIYQDVDKHYLSTTWLNDLQQEVKFDMYAYSSGGDDSKYPVTLKISDSQLFVSAQGEDQPVLLKELPETPKLITGSETDLIFFWKSINSKNYFTSAAYPELFIATKEQSRVHLARGLPSMTDFQIS
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1EU/μg) (determined by LAL gel clot method)
Gene Alias Il-1a
Common Name Il1a
Gene Symbol Il1a
Biological Activity The ED50 was determined by a cell proliferation assay using the murine helper T cell line, D10.G4., was found to be 10.0 pg/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.