Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Il20 (Q9JKV9, 152 amino acids) Partial Recombinant Protein

Catalog No. 89948802 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-802 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-802 Supplier Abnova™ Supplier No. P4474

Used for Func, SDS-PAGE

Specifications

Accession Number Q9JKV9
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 58181
Molecular Weight (g/mol) 17.6kDa
Name Il20 (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias MGC130536/Zcyto10
Common Name Il20
Gene Symbol Il20
Biological Activity The biological activity was determined by measuring the dose-dependent activation of STAT3 in human epidermal keratinocytes (HEK2a cells). Significant STAT3 activation is observed with ≥ 100ng/mL of recombinant mouse IL-20.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.