Learn More
Proteintech Group Inc Mouse IL21 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE1001296T
Encompass_Preferred
Interleukin 21 (IL21) is a cytokine produced predominantly by cluster of differentiation 4 (CD4+) T-cells and natural killer T-cells. IL21 is preliminarily expressed in activated cluster of differentiation 4 (CD4+) T-cells and natural killer (NK) T-cells. IL21 receptor (il21R) is a class I cytokine receptor, which requires dimerization with the indispensable common gamma chain (γc) subunit to bind IL21. IL21 plays a role in both the innate and adaptive immune responses by inducing the differentiation, proliferation and activity of multiple target cells including macrophages, natural killer cells, B cells and cytotoxic T cells. IL21 mediates apoptosis in B-CLL cells through up-regulation of the BIM (BH3 family member) and enhances both direct and antibody-dependent cellular cytotoxicity in these cells.
Catalog No. 50-197-4137
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.