Learn More
Sino Biological Mouse IL21R PAb 100UL 51184-T16-100
Supplier:  Sino Biological 51184T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-21R (rM IL-21R; Catalog#51184-M08H; AAL82636.1; Met1-Pro236). IL-21R specific IgG was purified by Mouse IL-21R affinity chromatography.
Catalog No. 50-240-8167
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.