Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Il7 (P10168) Recombinant Protein

Catalog No. 89948952 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-952 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-952 Supplier Abnova™ Supplier No. P4583
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Sequence: MECHIKDKEGKAYESVLMISIDELDKMTGTDSNCPNNEPNFFRKHVCDDTKEAAFLNRAARKLKQFLKMNISEEFNVHLLTVSQGTQTLVNCTSKEEKNVKEQKKNDACFLKRLLREIKTCWNKILKGSI

Specifications

Accession Number P10168
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 16196
Molecular Weight (g/mol) 17.4kDa
Name Il7 (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Immunogen MECHIKDKEGKAYESVLMISIDELDKMTGTDSNCPNNEPNFFRKHVCDDTKEAAFLNRAARKLKQFLKMNISEEFNVHLLTVSQGTQTLVNCTSKEEKNVKEQKKNDACFLKRLLREIKTCWNKILKGSI
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias A630026I06Rik/Il-7/MGC129342/hlb368
Common Name Il7
Gene Symbol Il7
Biological Activity The activity is determined by dose-dependent induction of 2E8 cell proliferation and is typically less than 0.2ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.