Learn More
Sino Biological Mouse IL7R MAb 100UL 50090-R115-100
Supplier:  Sino Biological 50090R115100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD127 / IL-7RA (rM CD127 / IL-7RA; Catalog#50090-M08H; NP_032398.3; Met1-Asp239).
Catalog No. 50-241-1857
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.