Learn More
ACROBiosystems ACROBiosystems Mouse Integrin alpha 10 beta 1 (ITGA10&ITGB1) Heterodimer Protein, His Tag&Tag Free
Supplier:  ACROBiosystems IT1M52W41MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse ITGA10&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-M52W4) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Thr 1118 (ITGA10) & Gln 21 - Asp 728 (ITGB1) (Accession # E9PXZ3-1 (ITGA10) & P09055-1 (ITGB1)).;Mouse ITGA10&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA10 and ITGB1, has a calculated MW of 126.6 kDa (ITGA10) and 83.5 kDa (ITGB1). Subunit ITGA10 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 150-170 kDa (ITGA10) and 100-130 kDa (ITGB1) respectively due to glycosylation.;Immobilized Human Collagen IV at 2 μg/mL (100 μL/well) can bind Mouse ITGA10&ITGB1 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT1-M52W4) with a linear range of 0.039-1.25 μg/mL (QC tested).
Catalog No. 50-210-7125
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.