Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Integrin beta 4/CD104 (NP_598424) VersaClone cDNA
SDP

Catalog No. RDC1966 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC1966 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC1966 Supplier R&D Systems™ Supplier No. RDC1966
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Mouse Integrin beta 4/CD104 (NP_598424) VersaClone cDNA is an integral cell-surface protein known to participate in cell adhesion as well as cell-surface mediated signaling.

The Mouse Integrin beta 4/CD104 (NP_598424) VersaClone cDNA clone contains the complete ORF for Integrin beta 4/CD104, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Integrin beta 4/CD104 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Specifications

Gene mItgb4
Accession Number NP_598424
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 5272bp
pH 8.5
Gene ID (Entrez) 3691
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.