Learn More
Biomatik Corporation Mouse Interleukin 24 (IL24) CLIA Kit
Supplier:  Biomatik Corporation EKU09880
Encompass_Preferred
Mouse Interleukin 24 (IL24) CLIA Kit for detecting IL24 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.59pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q925S4. Gene ID: 93672. Target Synonyms: C49A; FISP; IL10B; MDA7; Mob-5; ST16; Mda-7; Melanoma Differentiation Association Protein 7; Suppression Of Tumorigenicity 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4819
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.