Learn More
Description
- Also known as: JCAM; Jam4; 2010003D20Rik
- IGSF5 is member of the CTX (Cortical Thymocyte Marker in Xenopus) subfamily of the Ig superfamily.
- IGSF5 is expressed in intestinal, renal, and mammary epithelial cells, spermatogonia, and hematopoietic stem cells.
- It is a cell adhesion molecule that mediates homophilic interaction.
- IGSF5 also interacts with the intracellular membrane associated guanylate kinase inverted 1 (MAGI 1).
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mIgsf5 |
| Accession Number | AAP49218 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1126 bp |
| Gene ID (Entrez) | 150084 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
