Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Ki67/MKI67 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7649
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7649 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7649 Supplier R&D Systems Supplier No. AF7649
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 5 publications

Ki67/MKI67 Polyclonal specifically detects Ki67/MKI67 in Mouse samples. It is validated for Immunocytochemistry.

Specifications

Antigen Ki67/MKI67
Applications Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 5-15 ug/mL
Gene Accession No. NP_001074586
Gene Alias antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, Ki67, Ki-67, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, MKI67, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105, TSG126
Gene Symbols MKI67
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Ki-67/MKI67 Asn3053-Ser3177 Accession # NP_001074586
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4288
Test Specificity Detects mouse Ki-67/MKI67 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human Ki-67/MKI67 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.