Learn More
Biomatik Corporation Mouse Kidney Injury Molecule 1 (Kim1) CLIA Kit
Supplier:  Biomatik Corporation EKU08916
Encompass_Preferred
Mouse Kidney Injury Molecule 1 (Kim1) CLIA Kit for detecting Kim1 in tissue homogenates, urine, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 68.6-50,000pg/ml. Sensitivity: 26.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q5QNS5. Gene ID: 171283. Target Synonyms: HAVCR1; TIM1; TIMD1; HAVCR; Hepatitis A Virus Cellular Receptor 1; T Cell Immunoglobulin And Mucin Domain-Containing Protein 1; T-cell immunoglobulin mucin receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6095
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.