Learn More
Sino Biological Mouse KITLG PAb 100UL 50487-RP02-100
Supplier:  Sino Biological 50487RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse SCF / C-kit ligand (rM SCF / C-kit ligand; Catalog#50487-M08B; P20826-1; Met 1-Ala 189). SCF / C-kit ligand specific IgG was purified by Mouse SCF / C-kit ligand affinity chromatography.
Catalog No. 50-240-7684
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.