Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Leptin DuoSet ELISA
SDP

Catalog No. DY49805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY49805 1 Kit (5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY49805 Supplier R&D Systems™ Supplier No. DY49805
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Leptin DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Leptin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Leptin/OB.

Specifications

Assay Range 125.00 - 8000pg/mL
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 3952
Gene Symbol Lep
Quantity 1 Kit (5 Plates)
Sample Volume 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.