Learn More
Sartorius Incucyte Mouse IgG1 Fabfluor-pH Antibody Labeling Dye for Antibody Internalization, 50ug vial
Catalog No. NC1417772 Shop All Sartorius Products
Encompass
One vial (50 µg) of lyophilized conjugate sufficient for labeling large quantities of mouse (IgG1) test antibodies in 96-well format.  Incucyte Mouse IgG1 Fabfluor-pH Red Antibody Labeling Dye utilizes a novel pH-sensitive dye coupled antibody fragment, which labels Fc-containing mouse IgG1 test antibodies and allows for rapid antibody internalization and kinetic high-throughput analysis.
Catalog No. NC1417772 Supplier Sartorius Supplier No. 4723
One vial (50 µg) of lyophilized conjugate sufficient for labeling large quantities of mouse (IgG1) test antibodies in 96-well format.  Incucyte Mouse IgG1 Fabfluor-pH Red Antibody Labeling Dye utilizes a novel pH-sensitive dye coupled antibody fragment, which labels Fc-containing mouse IgG1 test antibodies and allows for rapid antibody internalization and kinetic high-throughput analysis.