Learn More
Description
- Lilra5 leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5 [ Mus musculus (house mouse) ]
- Also known as: Gm4878; Lilrc1; EG232801; E030017K20
- The LILRC1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- LILRA5/LILRC1 is a member of the leukocyte immunoglobulin-like receptor (LIR) family.
- LIR family members are known to have activating and inhibitory functions in leukocytes.
- LILRA5 is expressed on monocytes and perhaps neutrophils, and its activation results in the secretion of proinflammatory cytokines such as TNFa and IL 1b.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mLilra5 |
| Accession Number | NP_001074708 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 901 bp |
| Gene ID (Entrez) | 232801 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
