Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse LIX (CXCL6) (70 aa) Recombinant Protein, PeproTech®

Catalog No. 25036500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25036500UG 500 μg
25036100UG 100 μg
250-36-1MG 1 mg
250-36-20UG 20 μg
25036250UG 250 μg
250-36-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 25036500UG Supplier Gibco™ Supplier No. 25036500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine LIX is a 7.6 kDa protein consisting of amino acid residues 9-78 of the full length LIX protein (70 amino acids). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL6 is a chemotactic for neutrophil granulocytes. It signals through binding and activation of its receptors (CXCR1 and CXCR2). In addition to its chemotactic and angiogenic properties, it has strong antibacterial activity against Gram-positive and Gram-negative bacteria (90-fold-higher when compared to CXCL5 and CXCL7).
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Formulation protein with no preservative
Molecular Weight (g/mol) 7.6 kDa
Name Mouse LIX (CXCL6) (70 aa)
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); chemokine alpha 3; CKA3; CKA-3; CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine 6; C-X-C motif chemokine ligand 6; CXCL; Cxcl5; CXCL6; Cytokine LIX; GCP2; GCP-2; granulocyte chemotactic protein 2; Scyb5; SCYB6; small inducible cytokine subfamily B (Cys-X-Cys), member 6 (granulocyte chemotactic protein 2); Small inducible cytokine subfamily B (Cys-X-Cys), member b; small-inducible cytokine B5; Small-inducible cytokine B6; Small-inducible cytokine B6, N-processed variant 1; Small-inducible cytokine B6, N-processed variant 2; Small-inducible cytokine B6, N-processed variant 3
Common Name CXCL6
Conjugate Unconjugated
Recombinant Recombinant
Sequence TELRCVCLTV TPKINPKLIA NLEVIPAGPQ CPTVEVIAKL KNQKEVCLDP EAPVIKKIIQ KILGSDKKKA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.