Learn More
Sino Biological Mouse LRRC15 Protein 20UG 5A4998-M49H-B-20
Supplier:  Sino Biological 5A4998M49HB20
Encompass_Preferred
A DNA sequence encoding the Mouse LRRC15 (NP_083249.1) (Met1-Gly536) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1858
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.