Learn More
Description
- Tissue: Lung
- Host: Mouse
- Lysis Buffer: Modified RIPA Lysis Buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
- Storage buffer: In ready-to-use 1X Sample Buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blotting
Specifications
Specifications
| Description | Whole tissue lysate (denatured) |
| Lysis Buffer | Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. ;Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
| Preparation Method | Tissue lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). The lysate was adjusted to 3.75 mg/ml, and then mixed with 5X Sample Buffer to become final 3 mg/ml in 1X Sample Buffer. The lysate was heated at 95°C for 5 min, and cooled rapidly. |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 200 μg |
| Storage Buffer | In ready-to-use 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue). |
| Host Species | Mouse |
| Content And Storage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| For Use With (Application) | Western Blot |
| Tissue | Lung |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.