Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Lypla1 (NP_032892, 1 a.a. - 230 a.a.) Full-length Recombinant Protein with His tag

Catalog No. 89951312 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-951-312 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-951-312 Supplier Abnova™ Supplier No. P4342
Add to Cart
Add to Cart

Used for SDS-PAGE

Sequence: MGSSHHHHHHSSGLVPRGSHMCGNNMSAPMPAVVPAARKATAAVIFLHGLGDTGHGWAEAFAGIKSPHIKYICPHAPVMPVTLNMNMAMPSWFDIVGLSPDSQEDESGIKQAAETVKALIDQEVKNGIPSNRIILGGFSQGGALSLYTALTTQQKLAGVTALSCWLPLRASFSQGPINSANRDISVLQCHGDCDPLVPLMFGSLTVERLKALINPANVTFKIYEGMMHSSCQQEMMDVKHFIDKLLPPID

Specifications

Accession Number NP_032892
Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 18777
Molecular Weight (g/mol) 26.8kDa
Name Lypla1 (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing 15% SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMCGNNMSAPMPAVVPAARKATAAVIFLHGLGDTGHGWAEAFAGIKSPHIKYICPHAPVMPVTLNMNMAMPSWFDIVGLSPDSQEDESGIKQAAETVKALIDQEVKNGIPSNRIILGGFSQGGALSLYTALTTQQKLAGVTALSCWLPLRASFSQGPINSANRDISVLQCHGDCDPLVPLMFGSLTVERLKALINPANVTFKIYEGMMHSSCQQEMMDVKHFIDKLLPPID
Storage Requirements Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias Pla1a
Common Name Lypla1
Gene Symbol Lypla1
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.