Learn More
Description
- Also known as: rrg; TSC-160; AI893619
- LOX belongs to the lysyl oxidase family of proteins.
- The secreted proprotein is proteolytically processed to an active mature peptide and a propeptide.
- This propeptide is thought to function in tumor suppression by inhibiting the Ras signaling pathway.
- The active enzyme plays a role in cross-linking of collagen and elastin and is essential for development of cardiovascular and respiratory systems, and development of skin and connective tissue.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mLox |
| Accession Number | NP_034858 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1249 bp |
| Gene ID (Entrez) | 4015 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
