Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse M-CSF Recombinant Protein

Catalog No. PIRP8615
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8615 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8615 Supplier Invitrogen™ Supplier No. RP8615

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Recombinant Protein

Macrophage-CSF (mouse) Recombinant Protein, RP-8615, from Invitrogen.

M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells. Recombinant mouse M-CSF is a disulfide-linked homodimer, containing two 156 amino acid chains, with a total molecular weight of 36.4 kDa. Purity is typically greater than 98% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the dose-dependent induction of M-NFS-60 cell proliferation and is typically less than 2ng/mL. The specific activity of this protein is 5x10^5 units/mg. Reconstitute using sterile water at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07141
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation PBS with no preservative; pH 7.5
Gene ID (Entrez) 12977
Molecular Weight (g/mol) ∽36.8
Name Mouse M-CSF
pH Range 7.5
Quantity 10 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias M-MCSF
Common Name Mouse M-CSF
Gene Symbol Csf1
Specific Reactivity Mouse
Conjugate Unconjugated
Cross Reactivity Ms
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.