Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Mast Cell Protease-6/Mcpt6 ELISA Kit

Catalog No. EM51RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM51RB 96 Tests
EM51RBX5 5 x 96 Tests
EM51RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EM51RB Supplier Invitrogen™ Supplier No. EM51RB
Add to Cart
Add to Cart

ELISA

Mouse Mast Cell Protease-6/Mcpt6 quantitates mouse Mast Cell Protease-6/Mcpt6 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Mast Cell Protease-6/Mcpt6.

TPSB2 is a protein coding gene. Tryptases comprise a family of trypsin-like serine proteases, the peptidase family S1. Tryptases are enzymatically active only as heparin-stabilized tetramers, and they are resistant to all known endogenous proteinase inhibitors. Several tryptase genes are clustered on chromosome 16p13.3. These genes are characterized by several distinct features. They have a highly conserved 3' UTR and contain tandem repeat sequences at the 5' flank and 3' UTR which are thought to play a role in regulation of the mRNA stability. These genes have an intron immediately upstream of the initiator Met codon, which separates the site of transcription initiation from protein coding sequence. This feature is characteristic of tryptases but is unusual in other genes. The alleles of this gene exhibit an unusual amount of sequence variation, such that the alleles were once thought to represent two separate genes, beta II and beta III. Beta tryptases appear to be the main isoenzymes expressed in mast cells, whereas in basophils, alpha-tryptases predominate. Tryptases have been implicated as mediators in the pathogenesis of asthma and other allergic and inflammatory disorders.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AV011504; EC 3.4.21; EC 3.4.21.59; mast cell protease 6; mast cell tryptase beta II; mast cell tryptase beta III; Mcp6; Mcp-6; Mcpt6; MMCP-6; rMCP-6; testicular tissue protein Li 163; TPS2; TPS2tryptase beta-2; Tpsb2; tryptase beta 2; tryptase beta 2 (gene/pseudogene); tryptase beta II; tryptase beta III; tryptase beta-2; Tryptase II; tryptase III; Tryptase-2; tryptaseB; tryptaseC; Tryptase-C
Assay Range 0.84 to 200 ng/mL
Assay Sensitivity 0.84 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 17229
Gene Symbol Tpsb2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TPSB2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.