Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse MCP-5 (CCL12) Recombinant Protein, PeproTech®

Catalog No. 25004100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25004100UG 100 μg
250-04-1MG 1 mg
250-04-20UG 20 μg
25004250UG 250 μg
25004500UG 500 μg
250-04-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 25004100UG Supplier Gibco™ Supplier No. 25004100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine MCP-5 is a 9.3 kDa protein containing 82 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Chemotactic factor that attracts eosinophils, monocytes, and lymphocytes but not neutrophils. Potent monocyte active chemokine that signals through CCR2. Involved in allergic inflammation and the host response to pathogens and may play a pivotal role during early stages of allergic lung inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q62401
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 20293
Molecular Weight (g/mol) 9.3 kDa
Name Mouse MCP-5 (CCL12)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-C motif chemokine 12; Ccl12; chemokine (C-C motif) ligand 12; MCP-1-related chemokine; Mcp5; MCP-5; Monocyte chemoattractant protein 5; monocyte chemotactic protein 5; Scya12; small inducible cytokine A12; small-inducible cytokine A12
Common Name MCP-5
Gene Symbol Ccl12
Biological Activity Determined by its ability to chemoattract human peripheral blood monocytes using a concentration range of 10.0-50.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence GPDAVSTPVT CCYNVVKQKI HVRKLKSYRR ITSSQCPREA VIFRTILDKE ICADPKEKWV KNSINHLDKT SQTFILEPSC LG
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.