Learn More
ILEX LIFE SCIENCES LLC Ilex Life Sciences Mouse Macrophage Colony-Stimulating Factor 1 (M-CSF), E. coli Recombinant Protein, Bioactive, 10 ug
SDP
Supplier:  ILEX LIFE SCIENCES LLC A3439010
Encompass_Preferred
Our bioactive Macrophage Colony-Stimulating Factor 1 (M-CSF) Mouse, E. coli Recombinant Protein is a disulfide linked homodimer, non-glycosylated, polypeptide chain containing 2 x 156 amino acids and has a total molecular mass of 36.4 kDa. Purity: > 95%. Biological Activity: The ED50, as calculated by the dose-dependant stimulation of the proliferation of murine M-NFS-60 indicator cells is 1.33 ng/ml corresponding to a specific activity of 7.5x105 units/mg. Formulation: lyophilized from a sterile (0.2 µm) filtered solution containing 10mM sodium phosphate, 50mM sodium chloride, pH 7.5. Synonyms: Murine M-CSF, CSF-1, CSF1 gene product, MCSF. Summary: M-CSF is a cytokine that plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. M-CSF plays a role in immunological defenses, bone metabolism, lipoproteins clearance, fertility and pregnancy.
Catalog No. 50-211-9973
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.