Learn More
R&D Systems™ Mouse Mesenchymal Stem Cell Multi-Color Flow Kit
Catalog No. FMC003
Change view
Click to view available options
25 Tests
Catalog No.
FMC003 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. FMC003 Supplier R&D Systems™ Supplier No. FMC003
Add to Cart
Add to Cart
This item is not returnable. View return policy
For flow cytometric analysis of mouse MSCs using four fluorochrome-conjugated antibodies
Variations in techniques as well as differences in the MSC starting population may account for experimental variability and some of the contradictory data in the literature. By clearly defining the starting cell populations, researchers can decrease experimental variability and ultimately decrease apparent discrepancies in the literature that stem from the use of phenotypically different cell populations.
Verifies MSC identity in less than 2 hours
Included conjugated antibodies allow for single step staining
Defines the starting cell population
Multiple markers reduce experimental variation
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.