Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Meteorin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7065653
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB34752SP 25 μg
MAB34752 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB34752SP Supplier R&D Systems Supplier No. MAB34752SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Meteorin Monoclonal specifically detects Meteorin in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Meteorin
Applications ELISA
Classification Monoclonal
Clone 347511
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8C1Q4
Gene Alias C16orf23, c380A1.2, METRN
Gene Symbols Metrn
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Meteorin Gly22-Asp291 Accession # Q8C1Q4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79006
Test Specificity Detects mouse Meteorin in ELISAs.In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse CDNF, recombinant rat (rr) CNTF, or rrGDNF is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.