Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Mouse MFG-E8 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse MFG-E8 in cell culture supernates (25 ul), serum (10 ul), heparin plasma (10 ul).
Specifications
Specifications
| Assay Range | 0.13 - 8ng/mL |
| Assay Sensitivity | 0.02ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Mouse MFG-E8 |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 17304 |
| Quantity | 1 x 96-well Plate |
| Sample Volume | Cell Culture Supernates (25uL),Serum (10uL),Heparin Plasma (10uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
