Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Mouse MHC Class I ELISA Kit (Chemiluminescence)
Shop All Novus Biologicals Products

Specifications
Specifications
| Assay Range | 3.13 to 200 pg/mL |
| Assay Sensitivity | 1.88 pg/mL |
| Detection Method | Chemiluminescence |
| Sample Type | Serum, Plasma, Other Biological Fluids |
| Specificity | This kit recognizes Mouse MHC Class I in samples. No significant cross-reactivity or interference between Mouse MHC Class I and analogues was observed |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3133 |
| Gene Symbol | HLA-E |
| Quantity | 1 Kit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?