Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse MIP-1 gamma (CCL9/10) Recombinant Protein, PeproTech®

Catalog No. 25012250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
250 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25012250UG 250 μg
25012100UG 100 μg
250-12-1MG 1 mg
25012500UG 500 μg
250-12-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 25012250UG Supplier Gibco™ Supplier No. 25012250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine MIP-1gamma is an 11.6 kDa protein containing 101 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

MIP-1gamma is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1gamma signals through the CCR1 receptor. MIP-1gamma chemoattracts neutrophils, and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1gamma contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51670
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 20308
Molecular Weight (g/mol) 11.6 kDa
Name Mouse MIP-1 gamma (CCL9/10)
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-C motif chemokine 9; CCF18; Ccl9; CCL9(29-101); CCL9(30-101); CCL9(31-101); chemokine (C-C motif) ligand 9; chemokine CCL9; macrophage inflammatory protein 1-gamma; Macrophage inflammatory protein-related protein 2; MIP-1 gamma; MIP-1-gamma; Mrp2; MRP-2; RP23-320E6.3; Scya10; Scya9; small inducible cytokine A10; small inducible cytokine A9; Small-inducible cytokine A9
Common Name MIP-1 gamma
Gene Symbol Ccl9
Biological Activity Determined by its ability to chemoattract human neutrophils using a concentration range of 0.1-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQNSQPRTYK Q
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.