Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse MIS/AMH Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7064382
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1426SP 25 μg
MAB1426 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1426SP Supplier R&D Systems Supplier No. MAB1426SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

MIS/AMH Monoclonal specifically detects MIS/AMH in Mouse samples. It is validated for Western Blot.

Specifications

Antigen MIS/AMH
Applications Western Blot
Classification Monoclonal
Clone 188103
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P27106.1
Gene Alias AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance
Gene Symbols AMH
Host Species Rat
Immunogen E. coli-derived recombinant mouse MIS/AMH Asp450-Arg555 Accession # P27106.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268
Test Specificity Detects mouse MIS/AMH in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human MIS or recombinant rat MIS.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.