Learn More
ACROBiosystems HRP conjugated Anti-MMAE Antibody, mAb
Supplier:  ACROBiosystems MMEPLS104200UG
Encompass_Preferred
HRP conjugated Anti-MMAE Antibody, is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Bevacizumab MMAE.
Catalog No. 50-233-1452
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.