Learn More
Specifications
Specifications
| Gene | MMP-2 |
| Accession Number | NP_032636 |
| Gene ID (Entrez) | 4313 |
| Gene Alias | 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72 kDa gelatinase, 72 kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72 kD gelatinase, 72 kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP2, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1 matrix metalloproteinase 2 (gelatinase A, 72 kDa gelatinase, 72 kDa type IVcollagenase) |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
