Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse MMP3 ELISA Kit

Catalog No. EEL099
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL099 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL099 Supplier Invitrogen™ Supplier No. EEL099
Add to Cart
Edge
Add to Cart

ELISA

MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P28862
Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 17392
Gene Symbol MMP3
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target MMP3
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.