Learn More
American Research Products Inc Mouse myeloid differentiation
Supplier: American Research Products Inc CSBE12109M
Mouse myeloid differentiation factor 88 MyD88 ELISA Kit from CUSABIO Detection Range 47 pg/mL-3000 pg/mL Sensitivity 11 7 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.