Learn More
Stoelting™ Mouse and Neonatal Rat Adaptor
SDP
Catalog No. 10000171 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Mouse Posts, Neonatal Rat Adaptors
Mouse and Neonatal Rat Adaptors
Snout Clamp for Mouse Stereotaxics
Stereotaxic Ear Bars/Rubber Tips
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-171 Stereotaxic Ear Bars/Rubber Tips
10-000-152 Mouse and Neonatal Rat Adaptors
10-000-155 Snout Clamp for Mouse Stereotaxics
10-000-176 Mouse Posts, Neonatal Rat Adaptors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-000-171 Supplier Stoelting™ Supplier No. 51648
Only null left
Add to Cart
Add to Cart
Designed for use with traditional U-framed stereotaxic instruments like the Classic Lab Standard series
Model organisms with soft skulls, such as the mouse and the neonatal rat, are difficult to secure firmly in a traditional stereotaxic instrument. The stainless steel ear bars used for adult rats are too heavy to provide a secure hold without compressing the sides of the skull inward (which closes the airways and impedes blood fl ow). Further, the neonatal rat pup has no external opening of the auditory canal into which to insert the ear bars.
  • Ear and bite bar heights can be independently adjusted to accommodate a wide range of ages and assist in leveling the skull
  • Employs light, Delrin™ ear bars with tapered points on one end, and rubber tips on the other, to facilitate surgery on mice and rat pups
  • Unique feature permits ear bars to be independently adjusted in height to level the skull
  • Laser engraved scales show the vertical positions of the ear bars
  • Tooth bar and nose clamp secures the nose
  • Length accommodates larger, more mature animals
  • Specialized jaw holder cuffs securely clamp the zygomatic processes of the skull
  • Jaw holder cuffs fit over the ends of the ear bars, replacing the rubber tips, providing an alternative non-invasive means of securing the animal's head in the stereotaxic stage
  • Options for triple point securing of the animal's head in this adaptor make it exceptionally versatile
  • Set of two jaw holder cuffs included
Specifications
For Use With (Application) Neuroscience
Animal Rat
Model Stereotaxic Ear Bars/Rubber Tips