Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Netrin-1 Alexa Fluor Plus 594-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10141699
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10141699 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10141699 Supplier R&D Systems Supplier No. FAB1109AFP594100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Netrin-1 Monoclonal antibody specifically detects Netrin-1 in Mouse samples. It is validated for Western Blot,Gene Knock-Out

Specifications

Antigen Netrin-1
Applications Western Blot, Gene Knock-Out
Classification Monoclonal
Clone 158936
Conjugate Alexa Fluor Plus 594
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias netrin 1, netrin-1, Netrin1, NTN1, NTN1Lnetrin 1, mouse, homolog of
Host Species Rat
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9423
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8 °C as supplied
Product Type Antibody
Form Liquid
Isotype IgG2a
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.