Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Netrin-1 ELISA Kit

Catalog No. EM56RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM56RB 96 Tests
EM56RBX5 5 x 96 Tests
EM56RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EM56RB Supplier Invitrogen™ Supplier No. EM56RB
Add to Cart
Add to Cart

ELISA

Mouse Netrin-1 quantitates mouse Netrin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Netrin-1.

Netrin-1 is a protein target that plays a crucial role in various biological processes, particularly in the development and guidance of neuronal cells. It belongs to the netrin family of axon guidance molecules and is involved in mediating axon guidance, cell migration, and tissue morphogenesis during embryonic development. Netrin-1 functions by binding to specific receptors on the cell surface and providing directional cues for cellular movement and connectivity. Dysregulation of Netrin-1 has been implicated in various diseases, including cancer, neurological disorders, and cardiovascular conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AI561871; BC019633; Epididymis tissue protein Li 131 P; HGNC:8029; netrin 1; netrin 1, mouse, homolog of; Netrin1; netrin-1; Ntn1; NTN1L
Assay Range 0.09 to 20 ng/mL
Assay Sensitivity 0.09 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 18208
Gene Symbol NTN1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target NTN1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.