Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse NF-L ELISA Kit (Colorimetric)
SDP

Catalog No. NB013370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013370 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013370 Supplier Novus Biologicals™ Supplier No. NBP280299
Only null left
Add to Cart
Add to Cart

This kit recognizes Mouse NF in samples.

No significant cross-reactivity or interference between Mouse NF and analogues was observed.

Specifications

Assay Range 0.156 to 10 ng/mL
Assay Sensitivity 0.094 ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 4747
Gene Symbol NEFL
Host Species Mouse
Quantity 1 Kit
Research Discipline Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68 kDa
Target Neurofascin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.