Learn More
Description
- Also known as: Y4; Ppyr1; NYYR-D
- NPY4R is a member of the G protein-coupled receptor 1 family.
- It is one of five receptors for neuropeptide Y and peptide YY. NPY4R is expressed at high levels in amygdala, corpus callosum, hippocampus and subthalamic nucleus.
- Neuropeptide Y is one of the most abundant neuropeptides in the mammalian nervous system and exhibits a diverse range of important physiologic activities, including effects on psychomotor activity, food intake, regulation of central endocrine secretion, and potent vasoactive effects on the cardiovascular system.
- NPY receptors represent novel molecular therapeutic targets in cancer and obesity.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mNpy4r |
| Accession Number | NP_032945 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1141 bp |
| Gene ID (Entrez) | 5540 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
