Learn More
Biomatik Corporation Mouse Nuclear Factor Kappa B2 (NFkB2) ELISA Kit
Supplier:  Biomatik Corporation EKU06356
Encompass_Preferred
Mouse Nuclear Factor Kappa B2 (NFkB2) ELISA Kit for detecting NFkB2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTK5. Gene ID: 18034. Target Synonyms: LYT-10; H2TF1; Lymphocyte translocation chromosome 10; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 2 (p49/P100) ; Nuclear factor NF-kappa-B p52. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.