Learn More
Description
- Clec2d C-type lectin domain family 2, member d [ Mus musculus (house mouse) ]
- Also known as: Clrb; Ocil; Clr-b
- The OCIL/CLEC2d insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- CLEC2D is a type II transmembrane protein belonging to the natural killer cell receptor C-type lectin family.
- It is found on hematopoietic cells, osteoblasts, and chondrocytes.
- CLEC2D inhibits osteoclast formation and contains a transmembrane domain near the Nterminus as well as the C-type lectin-like extracellular domain.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mClec2d |
| Accession Number | NP_444339 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 637 bp |
| Gene ID (Entrez) | 29121 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
