Learn More
Description
- Osmr oncostatin M receptor [ Mus musculus (house mouse) ]
- Also known as: OSMRB
- The OSMR beta insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- OSMR is a member of the type I cytokine receptor family.
- It heterodimerizes with interleukin 6 signal transducer to form the type II oncostatin M receptor and with interleukin 31 receptor A to form the interleukin 31 receptor, and thus transduces oncostatin M and interleukin 31 induced signaling events.
- Mutations in OSMR have been associated with familial primary localized cutaneous amyloidosis.
Specifications
Specifications
| Gene | mOsmr |
| Accession Number | AAC40122 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2928 bp |
| Gene ID (Entrez) | 9180 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
